ENA245486621 Enamine

1-amino-3-(4-fluorophenoxy)propan-2-ol

  • MDL number MFCD07440124
  • Cas Number : 51448-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54317
Availability details for
Quantity
  • Availability
  • Add to Cart