ENA245486622 Enamine

{[5-(2-fluorophenyl)thiophen-2-yl]methyl}(propan-2-yl)amine

  • MDL number MFCD11629507
  • Cas Number : 1095117-78-4
  • Molecular Weight : 249.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-{[5-(2-fluorophenyl)-2-thienyl]methyl}-2-propanamine
Inchi Code 1S/C14H16FNS/c1-10(2)16-9-11-7-8-14(17-11)12-5-3-4-6-13(12)15/h3-8,10,16H,9H2,1-2H3
Inchi Key XQMDGFBBBFNLBS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54319
Availability details for
Quantity
  • Availability
  • Add to Cart