ENA245486625 Enamine

3-(difluoromethoxy)pyridine

  • MDL number MFCD13196036
  • Cas Number : 1209729-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54322
Availability details for
Quantity
  • Availability
  • Add to Cart