ENA245486629 Enamine

2-oxo-6-(pyridin-2-yl)-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00833383
  • Cas Number : 56304-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54331

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54331
Availability details for
Quantity
  • Availability
  • Add to Cart