ENA245486634 Enamine

5-(4-methoxyphenyl)pentanoic acid

  • MDL number MFCD00021791
  • Cas Number : 7508-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54340
Availability details for
Quantity
  • Availability
  • Add to Cart