ENA245486636 Enamine

2-amino-N-benzyl-N-ethylacetamide hydrochloride

  • MDL number MFCD13196039
  • Cas Number : 1209388-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54343

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54343
Availability details for
Quantity
  • Availability
  • Add to Cart