ENA245486638 Enamine

2-methyl-3-(morpholine-4-carbonyl)aniline

  • MDL number MFCD09043275
  • Cas Number : 926267-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53761
Availability details for
Quantity
  • Availability
  • Add to Cart