ENA245486640 Enamine

[4-(1,3-dioxaindan-5-yl)oxan-4-yl]methanamine

  • MDL number MFCD10691327
  • Cas Number : 1152568-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54349
Availability details for
Quantity
  • Availability
  • Add to Cart