ENA245486642 Enamine

2-[(2,5-difluorophenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD11523563
  • Cas Number : 162015-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54351
Availability details for
Quantity
  • Availability
  • Add to Cart