ENA245486645 Enamine

[1-(1,3-dioxaindan-5-yl)cyclohexyl]methanamine

  • MDL number MFCD10690216
  • Cas Number : 92500-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54354
Availability details for
Quantity
  • Availability
  • Add to Cart