ENA245486654 Enamine

cyclohexylmethanesulfonyl chloride

  • MDL number MFCD03452748
  • Cas Number : 4352-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53764

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53764
Availability details for
Quantity
  • Availability
  • Add to Cart