ENA245486657 Enamine

({4H,5H,6H-cyclopenta[b]thiophen-2-yl}methyl)(propan-2-yl)amine hydrochloride

  • MDL number MFCD13196050
  • Cas Number : 1211698-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54372
Availability details for
Quantity
  • Availability
  • Add to Cart