ENA245486662 Enamine

5-[(2,4-difluorophenyl)methyl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD13196053
  • Cas Number : 1209807-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-54377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54377
Availability details for
Quantity
  • Availability
  • Add to Cart