ENA245486663 Enamine

2-amino-2-benzyl-3-phenylpropanoic acid hydrobromide

  • MDL number MFCD13196054
  • Cas Number : 1211392-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54378
Availability details for
Quantity
  • Availability
  • Add to Cart