ENA245486665 Enamine

methyl 2-methyl-4-oxo-3,4-dihydroquinazoline-7-carboxylate

  • MDL number MFCD09815856
  • Cas Number : 1016681-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54380
Availability details for
Quantity
  • Availability
  • Add to Cart