ENA245486670 Enamine

1-methyl-4-[(piperidin-4-yl)methyl]piperazine

  • MDL number MFCD05215221
  • Cas Number : 735262-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54385
Availability details for
Quantity
  • Availability
  • Add to Cart