ENA245486671 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-methylbutanoic acid

  • MDL number MFCD09729578
  • Cas Number : 148991-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54386
Availability details for
Quantity
  • Availability
  • Add to Cart