ENA245486675 Enamine

3-cyclobutyl-1,2-oxazol-5-amine

  • MDL number MFCD11187392
  • Cas Number : 1039833-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54392
Availability details for
Quantity
  • Availability
  • Add to Cart