ENA245486682 Enamine

1-[4-(propan-2-yloxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD13195934
  • Cas Number : 103990-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53771

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53771
Availability details for
Quantity
  • Availability
  • Add to Cart