ENA245486683 Enamine

3-[(4-methylpiperazin-1-yl)sulfonyl]aniline

  • MDL number MFCD04035371
  • Cas Number : 436095-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-142
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54409
Availability details for
Quantity
  • Availability
  • Add to Cart