ENA245486691 Enamine

1-(4-chlorophenyl)-1-(thiophen-3-yl)methanamine hydrochloride

  • MDL number MFCD13196059
  • Cas Number : 1211615-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54421
Availability details for
Quantity
  • Availability
  • Add to Cart