ENA245486692 Enamine

4-amino-N-(4-fluorophenyl)benzene-1-sulfonamide

  • MDL number MFCD01016124
  • Cas Number : 1494-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54422
Availability details for
Quantity
  • Availability
  • Add to Cart