ENA245486694 Enamine

[2-(cyclopropylmethoxy)phenyl]methanol

  • MDL number MFCD11190320
  • Cas Number : 1039931-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54424
Availability details for
Quantity
  • Availability
  • Add to Cart