ENA245486701 Enamine

2-oxo-6-(pyridin-2-yl)-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD11841017
  • Cas Number : 56304-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 310...312
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54434
Availability details for
Quantity
  • Availability
  • Add to Cart