ENA245486702 Enamine

6-oxo-1,6-dihydro-[2,4'-bipyridine]-5-carbonitrile

  • MDL number MFCD00833385
  • Cas Number : 39883-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54435
Availability details for
Quantity
  • Availability
  • Add to Cart