ENA245486705 Enamine

2-{[3-(benzyloxy)phenyl]formamido}acetic acid

  • MDL number MFCD13196062
  • Cas Number : 1211741-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54438
Availability details for
Quantity
  • Availability
  • Add to Cart