ENA245486707 Enamine

3-[3-(trifluoromethyl)phenyl]-1,2-oxazol-5-amine

  • MDL number MFCD07787059
  • Cas Number : 82360-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54440
Availability details for
Quantity
  • Availability
  • Add to Cart