ENA245486710 Enamine

4-bromo-2-(propan-2-yloxy)benzoic acid

  • MDL number MFCD11651855
  • Cas Number : 1038595-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54443
Availability details for
Quantity
  • Availability
  • Add to Cart