ENA245486718 Enamine

2-methylpentanenitrile

  • MDL number MFCD09030401
  • Cas Number : 6339-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53774
Availability details for
Quantity
  • Availability
  • Add to Cart