ENA245486727 Enamine

N-cyclopropyl-N-ethyl-3,5-dimethyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD11618731
  • Cas Number : 1290685-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54472
Availability details for
Quantity
  • Availability
  • Add to Cart