ENA245486731 Enamine

N'-hydroxy-2-[(3-methoxyphenyl)methoxy]ethanimidamide

  • Cas Number : 1016864-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-54476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54476
Availability details for
Quantity
  • Availability
  • Add to Cart