ENA245486734 Enamine

4-(5-tert-butyl-1,2,4-oxadiazol-3-yl)aniline

  • MDL number MFCD12191342
  • Cas Number : 498547-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53782
Availability details for
Quantity
  • Availability
  • Add to Cart