ENA245486739 Enamine

3-(1-benzyl-1H-imidazol-2-yl)-4,4,4-trifluoro-3-hydroxybutanoic acid

  • MDL number MFCD13196077
  • Cas Number : 1197579-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54496

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54496
Availability details for
Quantity
  • Availability
  • Add to Cart