ENA245486742 Enamine

2-(1H-imidazol-2-yl)-1-(3-methylphenyl)ethan-1-one hydrochloride

  • MDL number MFCD13195940
  • Cas Number : 1197539-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53787
Availability details for
Quantity
  • Availability
  • Add to Cart