ENA245486744 Enamine

methyl 3-amino-3-(4-fluorophenyl)propanoate hydrochloride

  • MDL number MFCD13196082
  • Cas Number : 925412-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54503
Availability details for
Quantity
  • Availability
  • Add to Cart