ENA245486745 Enamine

1-[(1,3-thiazol-4-yl)methyl]piperazine

  • MDL number MFCD11170978
  • Cas Number : 933737-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53790
Availability details for
Quantity
  • Availability
  • Add to Cart