ENA245486750 Enamine

5-(4-fluoro-3-methylphenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD11155438
  • Cas Number : 1020969-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54509
Availability details for
Quantity
  • Availability
  • Add to Cart