ENA245486752 Enamine

(cyclobutylmethyl)thiourea

  • MDL number MFCD11634958
  • Cas Number : 1095505-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54511
Availability details for
Quantity
  • Availability
  • Add to Cart