ENA245486753 Enamine

3-{[4-(pyridin-2-yl)piperazin-1-yl]methyl}aniline

  • MDL number MFCD09804080
  • Cas Number : 1016726-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54512
Availability details for
Quantity
  • Availability
  • Add to Cart