ENA245486755 Enamine

4-(4-aminophenoxy)benzamide

  • MDL number MFCD11133127
  • Cas Number : 106273-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54514
Availability details for
Quantity
  • Availability
  • Add to Cart