ENA245486758 Enamine

2-[4-(4-aminophenoxy)phenyl]acetonitrile hydrochloride

  • MDL number MFCD13196085
  • Cas Number : 1211298-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-54517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54517
Availability details for
Quantity
  • Availability
  • Add to Cart