ENA245486760 Enamine

1-ethyl-1H-indol-5-amine

  • MDL number MFCD11213928
  • Cas Number : 220844-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54519

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54519
Availability details for
Quantity
  • Availability
  • Add to Cart