ENA245486763 Enamine

[(3-methylthiophen-2-yl)methyl](propan-2-yl)amine

  • MDL number MFCD09694047
  • Cas Number : 937667-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54522
Availability details for
Quantity
  • Availability
  • Add to Cart