ENA245486764 Enamine

5-(difluoromethyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD11193512
  • Cas Number : 25306-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53793

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53793
Availability details for
Quantity
  • Availability
  • Add to Cart