ENA245486768 Enamine

4-bromo-3-phenyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD13196088
  • Cas Number : 1238384-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260...262
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54550

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-54550
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart