ENA245486777 Enamine

2-(2-formylphenoxy)-N,N-dimethylacetamide

  • MDL number MFCD11149390
  • Cas Number : 124490-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54587

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54587
Availability details for
Quantity
  • Availability
  • Add to Cart