ENA245486778 Enamine

4H,5H,6H,7H-[1,2,4]triazolo[1,5-a]pyrimidine hydrochloride

  • MDL number MFCD13196091
  • Cas Number : 1208673-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54588

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-54588
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart