ENA245486780 Enamine

methyl 5-nitrothiophene-3-carboxylate

  • MDL number MFCD01934527
  • Cas Number : 88770-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54591

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54591
Availability details for
Quantity
  • Availability
  • Add to Cart