ENA245486783 Enamine

{1-[2-(dimethylamino)ethyl]piperidin-4-yl}methanamine

  • MDL number MFCD11131485
  • Cas Number : 1019381-31-7
  • Molecular Weight : 185.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54594
Availability details for
Quantity
  • Availability
  • Add to Cart