ENA245486785 Enamine

[(5-bromothiophen-3-yl)methyl](methyl)amine

  • MDL number MFCD12096200
  • Cas Number : 814255-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54596
Availability details for
Quantity
  • Availability
  • Add to Cart